missing translation for 'onlineSavingsMsg'
Learn More

FCER1A, Mouse, Clone: CRA1, Abnova™

Mouse monoclonal antibody raised against recombinant FCER1A.

Brand:  Abnova MAB7943.100ug

Product Code. 16101060

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The IgE receptor plays a central role in allergic disease, coupling allergen and mast cell to initiate the inflammatory and immediate hypersensitivity responses that are characteristic of disorders such as hay fever and asthma. The allergic response occurs when 2 or more high-affinity IgE receptors are crosslinked via IgE molecules that in turn are bound to an allergen (antigen) molecule. A perturbation occurs that brings about the release of histamine and proteases from the granules in the cytoplasm of the mast cell and leads to the synthesis of prostaglandins and leukotrienes--potent effectors of the hypersensitivity response. The IgE receptor consists of 3 subunits: alpha, beta (MIM 147138), and gamma (MIM 147139); only the alpha subunit is glycosylated.[supplied by OMIM

Specifications

Specifications

FCER1A
Monoclonal
Unconjugated
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
FCER1A
FCER1A
Recombinant protein corresponding to human FCER1A.
RUO
2205
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Flow Cytometry, Immunohistochemistry, Western Blot
CRA1
Mouse monoclonal antibody raised against recombinant FCER1A.
In PBS, pH 7.4 (50% glycerol)
FCE1A/FcERI
Mouse
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.