missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fcamr, Mouse, Clone: TX57, Abnova™
Mouse monoclonal antibody raised against native Fcamr.
Brand: Abnova MAB6584.50ug
This item is not returnable.
View return policy
Specifications
| Fcamr | |
| Monoclonal | |
| Unconjugated | |
| Immunoprecipitation (5-10 ug/mL) Flow Cyt (0.5-1 ug/mL) Immunoflourescence (5-10 ug/mL) Inhibitor Assay (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Fcamr | |
| Fcamr | |
| Native from Mouse Fcamr expressing cell line. | |
| RUO | |
| 64435 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Flow Cytometry, Immunoprecipitation, Inhibition Assays | |
| TX57 | |
| Mouse monoclonal antibody raised against native Fcamr. | |
| In PBS (0.1% proclin, 2% Block Ace) | |
| MGC129330/MGC129331 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction