Learn More
FBXO44, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FBXO44.
Brand: Abnova H00093611-A01.50uL
Beskrivning
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It is also a member of the NFB42 (neural F Box 42kDa) family, similar to F-box only protein 2 and F-box only protein 6. Four alternatively spliced transcript variants encoding two distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: PDCGSKYQLCVQLLSSAHAPLGTFQPDPATIQQKSDAKWREVSHTFSNYPPGVRYIWFQHGGVDTHYWAGWYGPRVTNSSITIGPPLPSpecifikationer
| FBXO44 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant FBXO44. | |
| FBXO44 | |
| DKFZp781J0852/FBG3/FBX30/FBX6A/Fbx44/Fbxo6a/MGC14140 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001014765 | |
| FBXO44 | |
| FBXO44 (NP_001014765, 168 a.a. to 255 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 93611 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only