missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-FANCD2, Clone: FI-17, Abnova™
Brand: Abnova MAB2398.100uL
This item is not returnable.
View return policy
Specifications
| Monoclonal | |
| Mouse monoclonal antibody raised against recombinant FANCD2. | |
| In buffer containing 0.09% sodium azide | |
| 100 μL | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| FI-17 | |
| ChIP (1:1000-1:2000) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| Recombinant fusion protein corresponding to FANCD2. | |
| Primary | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction