missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAH, Mouse, Clone: 26, Abnova™
Mouse monoclonal antibody raised against full-length recombinant FAH.
Brand: Abnova MAB9951.100ug
This item is not returnable.
View return policy
Description
This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT). [provided by RefSeq
Specifications
| FAH | |
| Monoclonal | |
| Unconjugated | |
| Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
| FAH | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 2184 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 26 | |
| Mouse monoclonal antibody raised against full length recombinant FAH. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| FAH | |
| Recombinant protein corresponding to full length human FAH. | |
| 100 μg | |
| Primary | |
| It can expression in HEK293 whole cell lysate. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction