missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAH, Mouse, Clone: 2, Abnova™
Description
This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT). [provided by RefSeq
Specifications
Specifications
| Antigen | FAH |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against full length recombinant FAH. |
| Dilution | Western blot (1:500) The optimal working dilution should be determined by the end user. |
| Formulation | In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) |
| Gene | FAH |
| Gene Symbols | FAH |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?