Learn More
FABP4, Mouse, Clone: 9B8D, Abnova™
Mouse monoclonal antibody raised against recombinant FABP4.
Brand: Abnova MAB10425.100uL
Description
FABP4 encodes the fatty acid binding protein found in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. [provided by RefSeq
Specifications
| FABP4 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| FABP4 | |
| FABP4 | |
| Recombinant protein corresponding to human FABP4. | |
| RUO | |
| 2167 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| 9B8D | |
| Mouse monoclonal antibody raised against recombinant FABP4. | |
| In ascitic (0.03% sodium azide) | |
| A-FABP/aP2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.