missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F5 Mouse anti-Bovine, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native F5.
Brand: Abnova MAB0260.100ug
This item is not returnable.
View return policy
Specifications
| F5 | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native F5. | |
| In PBS (50% glycerol) | |
| F5 | |
| Native purified Bovine F5. | |
| RUO | |
| 280687 | |
| Bovine | |
| Liquid |
| Radioimmune Assays (RIA), Western Blot | |
| Unconjugated | |
| Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| F5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody recognizes bovine factor Va light chain, bovine factor V in the absence of Ca2+. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction