missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/ERK2, Mouse, Clone: E31R, Abnova™
Mouse monoclonal antibody raised against recombinant ERK1/ERK2.
Brand: Abnova MAB9747.100ug
This item is not returnable.
View return policy
Specifications
| ERK1/ERK2 | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against recombinant ERK1/ERK2. | |
| In 10mM PBS, pH 7.2 (0.05% sodium azide) | |
| Recombinant protein corrsponding to full length human ERK1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| E31R | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein A purification | |
| RUO | |
| Recognizes human ERK1 and ERK2 proteins. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction