Learn More
EPHB1, Mouse, Clone: 5F10A4, Abnova™
Mouse monoclonal antibody raised against partial recombinant EPHB1.
Brand: Abnova MAB10595.100uL
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members. [provided by RefSeq
Specifications
| EPHB1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| EPHB1 | |
| EPHB1 | |
| Recombinant protein corresponding to amino acids 19-133 of human EPHB1. | |
| RUO | |
| 2047 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 5F10A4 | |
| Mouse monoclonal antibody raised against partial recombinant EPHB1. | |
| In ascites (0.03% sodium azide) | |
| ELK/EPHT2/FLJ37986/Hek6/NET | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.