Learn More
ELK1, Mouse, Clone: B4A12, Abnova™
Mouse monoclonal antibody raised against synthetic phosphopeptide of ELK1.
Brand: Abnova MAB6424.500ug
Description
This gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the the serum response factor and the serum reponse element in the promoter of the c-fos proto-oncogene. The protein encoded by this gene is a nuclear target for the ras-raf-MAPK signaling cascade. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| ELK1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| ELK1 | |
| Mouse | |
| 500 μg | |
| Primary | |
| Reactive to the immunogen and peptide-BSA conjugating. | |
| Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA | |
| B4A12 | |
| Mouse monoclonal antibody raised against synthetic phosphopeptide of ELK1. | |
| Lyophilized from 10mM PBS, pH 7.0 | |
| ELK1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S383 of human ELK1. | |
| RUO | |
| 2002 | |
| Human | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.