Learn More
EGFR, Mouse, Clone: M199, Abnova™
Mouse monoclonal antibody raised against synthetic phosphopeptide of EGFR.
Brand: Abnova MAB1382.100uL
Description
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq
Specifications
| EGFR | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:2000) Immunocytochemistry (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| EGFR | |
| EGFR | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y1101 of human EGFR. | |
| RUO | |
| 1956 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
| M199 | |
| Mouse monoclonal antibody raised against synthetic phosphopeptide of EGFR. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| ERBB/ERBB1/HER1/PIG61/mENA | |
| Mouse | |
| 100 μL | |
| Primary | |
| This human EGFR sequence has high homology to rat and mouse EGFR, and is not conserved in other ErbB family members. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.