missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBF1 (M01A), Mouse anti-Human, Clone: 1C12, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant EBF1.
Brand: Abnova H00001879-M01A.200uL
This item is not returnable.
View return policy
Description
Sequence: MFGIQESIQRSGSSMKEEPLGSGMNAVRTWMQGAGVLDANTAAQSGVGLARAHFEKQPPSNLRKSNFFHFVLALYDRQGQPVEIERTAFVGFVEKEKEANSEKTNNGIHYRLQLLYSNGIRTEQDFYVRLIDSMTKQAIVYEGQDKNPEMCRVLLTHEIMCSRCCDKKSCGNRNETPSDPVIIDRFFSKFFLKCNQNCLKNAGNPRDMRRFQVVVSTTVNVDGHVLAVSDNMFVHNNSKHGRRARRLDPSEGTPSYLEHATPCIKAISPSEGWTTGGATVIIIGDNFFDGLQVIFGTMLVWSELITPHAIRVQTPPRHIPGVVEVTLSYKSKQFCKGTPGRFIYTALNEPTIDYGFQRLQKVIPRHPGDPERLPKEVILKRAADLVEALYGMPHNNQEIILKRAADIAEALYSVPRNHNQLPALANTSVHAGMMGVNSFSGQLAVNVSEASQATNQGFTRNSSSVSPHGYVPSTTPQQTNYNSVTTSMNGYGSAAMSNLGGSPTFLNGSAANSPYAIVPSSPTMASSTSLPSNCSSSSGIFSFSPANMVSAVKQKSAFAPVVRPQTSPPPTCTSTNGNSLQAISGMIVPPMSpecifications
| EBF1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC038805 | |
| EBF1 | |
| EBF1 (AAH38805,1 a.a. ∽ 591 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1879 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1C12 | |
| Mouse monoclonal antibody raised against a full-length recombinant EBF1. | |
| EBF1 | |
| COE1/EBF/FLJ39389/FLJ41763/O/E-1/OLF1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction