Learn More
DSTN, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant DSTN.
Brand: Abnova H00011034-A01.50uL
Description
The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: LVGDVGVTITDPFKHFVGMLPEKDCRYALYDASFETKESRKEELMFFLWAPELAPLKSKMIYASSKDAIKKKFQGIKHECQANGPEDLNRACIAEKLGGSLIVAFEGCPSpecifications
| DSTN | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant DSTN. | |
| DSTN | |
| ACTDP/ADF/bA462D18.2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006870 | |
| DSTN | |
| DSTN (NP_006861, 56 a.a. to 164 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 11034 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only