missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drd5, Mouse, Clone: SG4-D1b, Abnova™
Mouse monoclonal antibody raised against partial recombinant Drd5.
Brand: Abnova MAB2313.100uL
This item is not returnable.
View return policy
Specifications
| Drd5 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Drd5 | |
| Mouse | |
| 100 μL | |
| Primary | |
| This antibody reacts with Dopamine Receptor D1B. This antibody is useful for Western Blot analysis, where a band is seen at 52-64 KDa in sf9 cell lysate. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| SG4-D1b | |
| Mouse monoclonal antibody raised against partial recombinant Drd5. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| Drd5 | |
| Recombinant protein corresponding to the C-terminus last 118 amino acids of rat Drd5. | |
| RUO | |
| 25195 | |
| Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction