Learn More
DPP4, Mouse, FITC, Clone: BA5b, Abnova™
Mouse monoclonal antibody raised against DPP4.
Brand: Abnova MAB9806.100Reactions
Description
The protein encoded by this gene is identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. [provided by RefSeq
Specifications
| DPP4 | |
| Monoclonal | |
| FITC | |
| Flow Cytometry (20 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user. | |
| DPP4 | |
| DPP4 | |
| A human T cell clone. | |
| RUO | |
| 1803 | |
| Human | |
| Liquid |
| Flow Cytometry | |
| BA5b | |
| Mouse monoclonal antibody raised against DPP4. | |
| In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide) | |
| ADABP/ADCP2/CD26/DPPIV/TP103 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| The mouse monoclonal antibody BA5b recognizes DPP4 (CD26), a 110 kDa type II membrane glycoprotein, which is a peptidase expressed on mature thymocytes, T cells (especially activated), B cells, NK cells and macrophages. | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.