missing translation for 'onlineSavingsMsg'
Learn More

DOC2A, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length recombinant DOC2A.

Brand:  Abnova H00008448-A01.50uL

Product Code. 16192462

  • £209.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

There are at least two protein isoforms of the Double C2 protein, namely alpha (DOC2A) and beta (DOC2B), which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2A is mainly expressed in brain and is suggested to be involved in Ca(2+)-dependent neurotransmitter release. [provided by RefSeq]

Sequence: MRGRRGDRMTINIQEHMAINVCPGPIRPIRQISDYFPRGPGPEGGGGSGGEAPAHLVPLALAPPAALLGATTPEDGAEVDSYDSDDATALGTLEFDLLYDRASCTLHCSILRAKGLKPMDFNGLADPYVKLHLLPGACKANKLKTKTQRNTLNPVWNEDLTYSGITDDDITHKVLRIAVCDEDKLSHNEFIGEIRVPLRRLKPSQKKHFNICLERQVPLASPSSMSAALRGISCYLKELEQAEQGQGLLEERGRILLSLSYSSRRRGLLVGILRCAHLAAMDVNGYSDPYVKTYLRPDVDKKSKHKTCVKKKTLNPEFNEEFFYEIELSTLATKTLEVTVWDYDIGKSNDFIGGVSLGPGARGEARKHWSDCLQQPDAALERWHTLTSELPPAAGALSSA
Specifications

Specifications

DOC2A
Polyclonal
Mouse polyclonal antibody raised against a full-length recombinant DOC2A.
DOC2A
DOC2A
DOC2A (AAH63436, 1 a.a. to 400 a.a) full-length recombinant protein with GST tag.
RUO
8448
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
50% glycerol
BC063436
Mouse
50 μL
Primary
Human
Antisera
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only