missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydroepiandrosterone (DHEA), Mouse, Clone: B133M, Abnova™
Mouse monoclonal antibody raised against Dehydroepiandrosterone (DHEA).
Brand: Abnova MAB3973.100ug
This item is not returnable.
View return policy
Specifications
| Dehydroepiandrosterone (DHEA) | |
| Monoclonal | |
| Unconjugated | |
| Recommend dilutions start at 5 ug/mL. The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| ELISA | |
| B133M | |
| Mouse monoclonal antibody raised against Dehydroepiandrosterone (DHEA). | |
| In PBS (0.09% sodium azide) | |
| Dehydroepiandrosterone (DHEA) | |
| RUO | |
| Reacts with DHEA (Dehydroepiandrosterone), DHEA- sulfate; <1% cross-reactivity with other steroids including cholesterol, progesterone, hydrocortisone, estradiol and Vitamin D3. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction