missing translation for 'onlineSavingsMsg'
Learn More

DEGS1, Mouse, Clone: 3F9, Abnova™

Mouse monoclonal antibody raised against a partial recombinant DEGS1.

Brand:  Abnova H00008560-M01A.200uL

Product Code. 16188825

  • £303.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. This protein contains three His-containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It is predicted to be a multiple membrane-spanning protein localized to the endoplasmic reticulum. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulating biosynthetic processing of the EGF receptor. Two splice variants have been identified. [provided by RefSeq

Sequence: SGHFIAEHYMFLKGHETYSYYGPLNLLTFNVGYHNEHHDFPNIPGKSLPLVRKIAAEYYDNLPHYNSWIKVLYDFVMDDTISPYSRMKRHQKGEMVLE
Specifications

Specifications

DEGS1
Monoclonal
Unconjugated
ascites with no preservative
NM_003676
DEGS1
DEGS1 (NP_003667,226 a.a. ∽ 323 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
8560
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Ascites
ELISA, Western Blot
3F9
Mouse monoclonal antibody raised against a partial recombinant DEGS1.
DEGS1
DEGS/DES1/Des-1/FADS7/MGC5079/MIG15/MLD
Mouse
200 μL
Primary
Human
Antibody
IgM κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.