Learn More
DEFA5, Mouse, Clone: 8C8, Abnova™
Mouse monoclonal antibody raised against full-length recombinant DEFA5.
Brand: Abnova MAB2329.100uL
Description
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several of the alpha defensin genes appear to be clustered on chromosome 8. The protein encoded by this gene, defensin, alpha 5, is highly expressed in the secretory granules of Paneth cells of the ileum. [provided by RefSeq
Specifications
| DEFA5 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| DEFA5 | |
| DEFA5 | |
| Recombinant protein corresponding to full length human DEFA5. | |
| RUO | |
| 1670 | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 8C8 | |
| Mouse monoclonal antibody raised against full length recombinant DEFA5. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| DEF5/HD-5/MGC129728 | |
| Mouse | |
| 100 μL | |
| Primary | |
| This antibody is specific to HD5. It does not cross-react with HD6, lysozyme or sPLA2. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.