Learn More
DDX39, Mouse, Clone: 2E4, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of DDX39.
Brand: Abnova MAB6576.50ug
Description
This gene encodes a member of the DEAD box protein family. These proteins are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. [provided by RefSeq
Specifications
| DDX39 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1 ug/mL) Immunohistochemistry (3 ug/mL) The optimal working dilution should be determined by the end user. | |
| DDX39 | |
| DDX39 | |
| A synthetic peptide corresponding to N-terminus of human DDX39. | |
| RUO | |
| 10212 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry | |
| 2E4 | |
| Mouse monoclonal antibody raised against synthetic peptide of DDX39. | |
| In PBS (0.1% proclin, 2% Block Ace) | |
| BAT1/BAT1L/DDXL/MGC18203/MGC8417/URH49 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.