missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer, Mouse, Clone: DD4, Abnova™
Mouse monoclonal antibody raised against D-Dimer.
Brand: Abnova MAB2921.200ug
This item is not returnable.
View return policy
Specifications
| D-Dimer | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| DD4 | |
| Mouse monoclonal antibody raised against D-Dimer. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Homogenized fibrin clot, human D-dimer or high molecular weight fibrin degradation products. | |
| RUO | |
| This antibody recognizes D-dimer and high molecular weight fibrin degradation products with different specificities. | |
| Store at 4°C. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction