Learn More
CYP4F11, Mouse, Clone: F21 P6 F5, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CYP4F11.
Brand: Abnova MAB1861.200ug
Description
This gene, CYP4F11, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 19. Another member of this family, CYP4F2, is approximately 16kb away. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| CYP4F11 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CYP4F11 | |
| Mouse | |
| 200 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| F21 P6 F5 | |
| Mouse monoclonal antibody raised against synthetic peptide of CYP4F11. | |
| In PBS (0.08% sodium azide) | |
| CYP4F11 | |
| A synthetic peptide corresponding to C-terminus of human CYP4F11. | |
| RUO | |
| 57834 | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.