Learn More
CYP3A7, Mouse, Clone: F19 P2 H2, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CYP3A7.
Brand: Abnova MAB1860.200ug
Description
This gene, CYP3A7, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme hydroxylates testosterone and dehydroepiandrosterone 3-sulphate, which is involved in the formation of estriol during pregnancy. The enzyme also metabolizes some drugs such as aflatoxin B1. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Transcript variants have been described, but it is not known whether these transcripts are normally produced. [provided by RefSeq
Specifications
| CYP3A7 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CYP3A7 | |
| CYP3A7 | |
| A synthetic peptide corresponding to C-terminus of human CYP3A7. | |
| RUO | |
| 1551 | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| F19 P2 H2 | |
| Mouse monoclonal antibody raised against synthetic peptide of CYP3A7. | |
| In PBS (0.08% sodium azide) | |
| CP37/P450-HFLA | |
| Mouse | |
| 200 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.