missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-CtBP1, Clone: 4D6, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74040-100UL
This item is not returnable.
View return policy
Description
CtBP1 Monoclonal specifically detects CtBP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CtBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, ELISA 1:100-1:2000 | |
| NP_001319 | |
| CTBP1 | |
| Recombinant human CTBP1 (1-440aa) purified from E. coli | |
| 100 μL | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 1487 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4D6 | |
| Unconjugated | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human CTBP1 mAb, clone 4D6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CTBP1 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction