Learn More
CSF3R, Mouse, Clone: S-1268, Abnova™
Mouse monoclonal antibody raised against recombinant CSF3R.
Brand: Abnova MAB6262.500ug
Description
The protein encoded by this gene is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble. Mutations in this gene are a cause of Kostmann syndrome, also known as severe congenital neutropenia. [provided by RefSeq
Specifications
| CSF3R | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CSF3R | |
| CSF3R | |
| Recombinant MBP fusion protein corresponding to human CSF3R. | |
| RUO | |
| 1441 | |
| Human | |
| Lyophilized |
| ELISA, Immunoprecipitation, Western Blot | |
| S-1268 | |
| Mouse monoclonal antibody raised against recombinant CSF3R. | |
| Lyophilized from 10mM PBS, pH 7.2 | |
| CD114/GCSFR | |
| Mouse | |
| 500 μg | |
| Primary | |
| This antibody is reactive with GSF3R and does not cross-react with MBP or BSA. | |
| Store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.