Learn More
CRKL, Mouse, Clone: 5E12, Abnova™
Mouse monoclonal antibody raised against partial recombinant CRKL.
Brand: Abnova MAB10613.100uL
Description
This gene encodes a protein kinase containing SH2 and SH3 (src homology) domains which has been shown to activate the RAS and JUN kinase signaling pathways and transform fibroblasts in a RAS-dependent fashion. It is a substrate of the BCR-ABL tyrosine kinase, plays a role in fibroblast transformation by BCR-ABL, and may be oncogenic
Specifications
| CRKL | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| CRKL | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Western Blot | |
| 5E12 | |
| Mouse monoclonal antibody raised against partial recombinant CRKL. | |
| In ascites (0.03% sodium azide) | |
| CRKL | |
| Recombinant protein corresponding to human CRKL. | |
| RUO | |
| 1399 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.