Learn More
CPB2 Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native CPB2.
Brand: Abnova MAB0274.100ug
Description
Carboxypeptidases are enzymes that hydrolyze C-terminal peptide bonds. The carboxypeptidase family includes metallo-, serine, and cysteine carboxypeptidases. According to their substrate specificity, these enzymes are referred to as carboxypeptidase A (cleaving aliphatic residues) or carboxypeptidase B (cleaving basic amino residues). The protein encoded by this gene is activated by trypsin and acts on carboxypeptidase B substrates. After thrombin activation, the mature protein downregulates fibrinolysis. Polymorphisms have been described for this gene and its promoter region. Available sequence data analyses indicate splice variants that encode different isoforms. [provided by RefSeq
Specifications
| CPB2 | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native CPB2. | |
| In PBS (50% glycerol) | |
| CPU/PCPB/TAFI | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody recognizes human TAFI, activated TAFI. Inhibits TAFI activation. It will inhibits the activated TAFI. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CPB2 | |
| CPB2 | |
| Native purified HumanCPB2. | |
| RUO | |
| 1361 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.