Learn More
COPA, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant COPA.
Marca: Abnova H00001314-A01.50uL
Descripción
In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by nonclathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms. [provided by RefSeq
Sequence: TKFETKSARVKGLSFHPKRPWILTSLHNGVIQLWDYRMCTLIDKFDEHDGPVRGIDFHKQQPLFVSGGDDYKIKVWNYKLRRCLFTLLGHLDYIRTTFEspecificaciones
| COPA | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant COPA. | |
| COPA | |
| FLJ26320/HEP-COP | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004371 | |
| COPA | |
| COPA (NP_004362, 3 a.a. to 100 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 1314 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only