Learn More
COL5A3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant COL5A3.
Brand: Abnova H00050509-A01
Description
This gene encodes an alpha chain for one of the low abundance fibrillar collagens. Fibrillar collagen molecules are trimers that can be composed of one or more types of alpha chains. Type V collagen is found in tissues containing type I collagen and appears to regulate the assembly of heterotypic fibers composed of both type I and type V collagen. This gene product is closely related to type XI collagen and it is possible that the collagen chains of types V and XI constitute a single collagen type with tissue-specific chain combinations. Mutations in this gene are thought to be responsible for the symptoms of a subset of patients with Ehlers-Danlos syndrome type III. Messages of several sizes can be detected in northern blots but sequence information cannot confirm the identity of the shorter messages. [provided by RefSeq
Sequence: EMVTLVADCEAQPPVLGHGPRFISIAGLTVLGTQDLGEKTFEGDIQELLISPDPQAAFQACERYLPDCDNLAPAATVAPQGEPETPRPSpecifications
| COL5A3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant COL5A3. | |
| COL5A3 | |
| COL5A3 | |
| COL5A3 (NP_056534, 157 a.a. to 244 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 50509 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_015719 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only