Learn More
CLEC4E, Mouse, Clone: AT16E3, Abnova™
Mouse monoclonal antibody raised against partial recombinant CLEC4E.
Brand: Abnova MAB8479.100uL
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type II transmembrane protein is a downstream target of CCAAT/enhancer binding protein (C/EBP), beta (CEBPB) and may play a role in inflammation. Alternative splice variants have been described but their full-length sequence has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region. [provided by RefSeq
Specifications
| CLEC4E | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:250) Western Blot (1:250-1:500) Immunofluorescence (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| CLEC4E | |
| CLEC4E | |
| Recombinant protein corresponding to amino acids 41-219 of human CLEC4E. | |
| RUO | |
| 26253 | |
| Store at 4°C short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| AT16E3 | |
| Mouse monoclonal antibody raised against partial recombinant CLEC4E. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CLECSF9/MINCLE | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.