missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED4, Mouse, Clone: HT13-2D6.3, Abnova™
Mouse monoclonal antibody raised against full-length recombinant CITED4.
Brand: Abnova MAB2352.100uL
This item is not returnable.
View return policy
Description
CITED4 belongs to a family of transcriptional coactivators that bind several proteins, including CREB-binding protein (MIM 600140) and p300 (MIM 602700).[supplied by OMIM
Specifications
| CITED4 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CITED4 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| HT13-2D6.3 | |
| Mouse monoclonal antibody raised against full length recombinant CITED4. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| CITED4 | |
| Recombinant fusion protein corresponding to full length human CITED4. | |
| RUO | |
| 163732 | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction