missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP7, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CHMP7 protein.
Brand: Abnova H00091782-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MWSPEREAEAPAGGDPAGLLPPEWEEDEERMSFLFSAFKRSREVNSTDWDSKMGFWAPLVLSHSRRQGVVRLRLRDLQEAFQRKGSVPLGLATVLQDLLRRGELQRESDFMASVDSSWISWGVGVFLLKPLKWTLSNMLGDNKVPAEEVLVAVELLKEKAEEVYRLYQNSPLSSHPVVALSELSTLCANSCPDERTFYLVLLQLQKEKRVTVLEQNGEKIVKFARGPRAKVSPVNDVDVGVYQLMQSEQLLSRKVESLSQEAERCKEEARRACRAGKKQLALRSLKAKQRTEKRIEALHAKLDTVQGILDRIYASQTDQMVFNAYQAGVGALKLSMKDVTVEKAESLVDQIQELCDTQDEVSQTLAGGVTNGLDFDSEELEKELDILLQDTTKEPLDLPDNPRNRHFTNSVPNPRISDAELEAELEKLSLSEGGLVPSSKSPKRQLEPTLKPLSpecifications
| CHMP7 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human CHMP7 protein. | |
| CHMP7 | |
| MGC29816 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 91782 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_152272.2 | |
| CHMP7 | |
| CHMP7 (NP_689485.1,1 a.a. ∽ 453 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction