missing translation for 'onlineSavingsMsg'
Learn More

CHIT1, Mouse, Clone: 1D9G2, 1D9G4, 2B4G3, Abnova™

Mouse monoclonal antibody raised against recombinant CHIT1.

Brand:  Abnova MAB10472.100uL

Product Code. 16060246

  • £528.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Chitotriosidase is secreted by activated human macrophages and is markedly elevated in plasma of Gaucher disease patients. The expression of chitotriosidase occurs only at a late stage of differentiation of monocytes to activated macrophages in culture. Human macrophages can synthesize a functional chitotriosidase, a highly conserved enzyme with a strongly regulated expression. This enzyme may play a role in the degradation of chitin-containing pathogens. [provided by RefSeq

Specifications

Specifications

CHIT1
Monoclonal
Unconjugated
Western Blot (1:500-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
CHIT1
CHIT1
Recombinant protein corresponding to human CHIT1.
RUO
1118
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG1
ELISA, Western Blot
1D9G2, 1D9G4, 2B4G3
Mouse monoclonal antibody raised against recombinant CHIT1.
In ascitic (0.03% sodium azide)
CHI3/CHIT/FLJ00314/MGC125322
Mouse
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.