Learn More
CGB, Mouse, Clone: hCG 205, Abnova™
Mouse monoclonal antibody raised against CGB.
Brand: Abnova MAB6189.500uL
Description
This gene is a member of the glycoprotein hormone beta chain family and encodes the beta 3 subunit of chorionic gonadotropin (CG). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. CG is produced by the trophoblastic cells of the placenta and stimulates the ovaries to synthesize the steroids that are essential for the maintenance of pregnancy. The beta subunit of CG is encoded by 6 genes which are arranged in tandem and inverted pairs on chromosome 19q13.3 and contiguous with the luteinizing hormone beta subunit gene. [provided by RefSeq
Specifications
| CGB | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| CGB | |
| CGB | |
| Human CGB. | |
| RUO | |
| 1082 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| hCG 205 | |
| Mouse monoclonal antibody raised against CGB. | |
| In PBS, pH 7.4 (1% BSA, 0.05% sodium azide) | |
| CGB3/hCGB | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.