missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFP, Mouse, Clone: 98s55, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CFP.
Brand: Abnova MAB8760.100ug
This item is not returnable.
View return policy
Specifications
| CFP | |
| Monoclonal | |
| 1 ug/uL | |
| Mouse monoclonal antibody raised against synthetic peptide of CFP. | |
| In PBS, pH 7.4 (0.02% sodium azide) | |
| A synthetic peptide (conjugated with carrier KLH) corresponding to N-terminus amino acids of Aequorea victoria CFP. | |
| 100 μg | |
| Primary | |
| Jellyfish | |
| Liquid |
| Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 98s55 | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Affinity purification | |
| RUO | |
| This antibody recognizes native and denatured forms of GFP and its variants EGFP, YFP, EYFP, and CFP. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction