Learn More
CER1, Mouse, Clone: 9D6, Abnova™
Mouse monoclonal antibody raised against recombinant CER1.
Brand: Abnova MAB10470.100uL
Description
This gene encodes a cytokine member of the cysteine knot superfamily, characterized by nine conserved cysteines and a cysteine knot region. The cerberus-related cytokines, together with Dan and DRM/Gremlin, represent a group of bone morphogenetic protein (BMP) antagonists that can bind directly to BMPs and inhibit their activity. [provided by RefSeq
Specifications
| CER1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| CER1 | |
| CER1 | |
| Recombinant protein corresponding to human CER1. | |
| RUO | |
| 9350 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 9D6 | |
| Mouse monoclonal antibody raised against recombinant CER1. | |
| In ascitic (0.03% sodium azide) | |
| DAND4/MGC119894/MGC119895/MGC96951 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.