missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5, Mouse, Clone: 1C7, Abnova™
Mouse monoclonal antibody raised against recombinant CEACAM5.
Brand: Abnova MAB10469.100uL
This item is not returnable.
View return policy
Specifications
| CEACAM5 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| CEACAM5 | |
| CEACAM5 | |
| Recombinant protein corresponding to human CEACAM5. | |
| RUO | |
| 1048 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 1C7 | |
| Mouse monoclonal antibody raised against recombinant CEACAM5. | |
| In ascitic (0.03% sodium azide) | |
| CD66e/CEA/DKFZp781M2392 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction