Learn More
CDH5, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against partial recombinant CDH5.
Brand: Abnova MAB8745.100ug
Description
This gene is a classical cadherin from the cadherin superfamily and is located in a six-cadherin cluster in a region on the long arm of chromosome 16 that is involved in loss of heterozygosity events in breast and prostate cancer. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, the protein may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but its full length sequence has not been determined. [provided by RefSeq
Specifications
| CDH5 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:100-1:2000) ELISA (1:5000-1:20000) The optimal working dilution should be determined by the end user. | |
| CDH5 | |
| 7B4/CD144/FLJ17376 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 1003 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against partial recombinant CDH5. | |
| In PBS, pH 7.4 (0.02% sodium azide, 1% BSA, 50% glycerol) | |
| NM_001795.3 | |
| CDH5 | |
| Recombinant protein corresponding to N-terminus residues of human CDH5. | |
| 100 μg | |
| Primary | |
| This antibody is specific to CDH5. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.