Learn More
CD92 Mouse anti-Human, Alexa Fluor 647, Clone: VIM15, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565316
Description
The VIM15 monoclonal antibody specifically binds to CD92 which is also known as Solute carrier family 44 member 1 (SLC44A1), or Choline transporter-like protein 1 (CHTL1). CD92 is a multipass membrane glycoprotein that belongs to the choline transporter-like family. It contains a tyrosine-based inhibitory motif (ITIM) in its cytoplasmic C-terminus. CD92 appears to be involved in choline transport for cell membrane phospholipid synthesis and in regulatory cell-signaling pathways. CD92 is strongly expressed by monocytes and at lower levels by granulocytes and some dendritic cells. It is generally expressed at low levels by B cells, endothelial cells, epithelial cells, and subsets of T cells, or NK cells. The VIM15 antibody has been shown to augment lipopolysaccharide-induced IL-10 production by monocyte-derived dendritic cells (Mo-DC). When cultured with ionomycin or calcium ionophore, maturing Mo-DC showed downregulation of CD92 expression.
Specifications
| CD92 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Myeloid Cell Line MV4-11 | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| VIM15 | |
| SLC44A1; Choline transporter-like protein 1; CHTL1; CTL1; RP11-287A8.1; p70 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.