missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80, Mouse, Clone: P1.H5.A1.A1, Abnova™
Mouse monoclonal antibody raised against CD80.
Brand: Abnova MAB6905.100ug
This item is not returnable.
View return policy
Description
The B-lymphocyte activation antigen B7-1 (formerly referred to as B7) provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 (MIM 186760) and CTLA4 (MIM 123890) ligands of T cells.[supplied by OMIM
Specifications
| CD80 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CD80 | |
| CD80 | |
| Human CD80 (B7-1) Ig Fusion Protein. | |
| RUO | |
| 941 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry | |
| P1.H5.A1.A1 | |
| Mouse monoclonal antibody raised against CD80. | |
| In 50mM sodium phosphate buffer, 100mM potassium chloride, 150mM NaCl, pH 7.5 (0.5mg/mL gentamicin sulfate) | |
| CD28LG/CD28LG1/LAB7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction