Learn More
CD56 Mouse anti-Human, APC-R700, Clone: NCAM16.2, BD
Mouse Monoclonal Antibody
£162.90 - £733.00
Specifications
| Antigen | CD56 |
|---|---|
| Clone | NCAM16.2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-R700 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15894109
|
BD Biosciences
565140 |
25 Tests |
£170.00 £162.90 / Each Save £7.10 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15884109
|
BD Biosciences
565139 |
100 Tests |
£733.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NCAM16.2 monoclonal antibody specifically binds to human CD56. It recognizes an extracellular immunoglobulin-like domain common to 120, 140, and 180 kDa forms of CD56, also known as the neural cell adhesion molecule (NCAM), NKH1 or MSK39. The CD56 antigen is expressed on approximately 10% to 25% of peripheral blood lymphocytes. It is present on essentially all resting and activated CD16+ natural killer (NK) lymphocytes and approximately 5% of CD3+ peripheral blood lymphocytes. CD3+ CD56+ T lymphocytes comprise a unique subset of cytotoxic T lymphocytes that mediates non-major histocompatibility complex (MHC)-restricted cytotoxicity. CD56 antigen density on NK lymphocytes increases upon cellular activation. The CD56 antigen is involved in neuronal homotypic cell adhesion and cell differentiation during embryogenesis. CD16+ CD56+ NK cells demonstrate reciprocal transfer of an activation state with dendritic cells.
This antibody was conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45-nm filter).
Specifications
| CD56 | |
| Flow Cytometry | |
| APC-R700 | |
| RUO | |
| Aqueous buffered solution containing BSA, protein stabilizer, glycerol, and ≤0.09% sodium azide. | |
| IgG2b κ | |
| Affinity Purified | |
| NCAM1; NCAM-1; NCAM; Leu-19; Neural cell adhesion molecule 1; NKH1; MSK39 |
| NCAM16.2 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Immunoaffinity-enriched adult human brain NCAM | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.