Learn More
CD48, Mouse, Biotin, Clone: MEM-102, Abnova™
Mouse monoclonal antibody raised against native CD48.
Brand: Abnova MAB4636.100ug
Description
BLAST1 is the designation used for an activation-associated cell surface glycoprotein of 40 to 45kDa expressed primarily in mitogen-stimulated human lymphocytes. The protein sequence predicted by the cDNA encoding BLAST1 indicates that BLAST1 is a member of the immunoglobulin supergene family. Yokoyama (1991) identified the BLAST1 activation/adhesion molecule as CD48.[supplied by OMIM
Specifications
| CD48 | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against native CD48. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| BCM1/BLAST/BLAST1/MEM-102/SLAMF2/hCD48/mCD48 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with CD48 antigen (Blast-1), a 40-47 KDa GPI-anchored membrane protein (immunoglobulin supergene family) widely expressed on hematopoietic cells; it is negative on granulocytes, platelets and erythrocytes. | |
| Store at 4°C. | |
| IgG1 |
| Flow Cytometry | |
| MEM-102 | |
| Biotin | |
| Flow Cytometry (1:1000) The optimal working dilution should be determined by the end user. | |
| CD48 | |
| CD48 | |
| Native purified CD48 from Raji Burkitt's lymphoma cell line. | |
| RUO | |
| 962 | |
| Human, Non-human Primate | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.