Learn More
CD266 (TWEAK-R) Mouse anti-Human, PE, Clone: ITEM-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565731
Description
The ITEM-1 monoclonal antibody specifically binds to CD266, which is also known as TWEAK Receptor (TWEAK-R/TWEAKR), or Fibroblast growth factor inducible 14 (FGF-inducible 14, FN14). CD266 is a ∽14 kDa type I transmembrane protein that is the 12A family member of the tumor necrosis factor receptor superfamily (TNFRSF12A). It is expressed at low levels in a variety of tissues including, heart, placenta, lung, muscle, and pancreas. CD266 is relatively highly expressed on HUVEC cells and certain tumor cell lines. TWEAK-induced CD266-mediated signaling can play roles in inflammation, the induction of apoptosis in certain cell types, the proliferation and migration of endothelial cells, and can promote angiogenesis within healthy and diseased tissues, eg, tumors. The ITEM-1 antibody can reportedly induce the proliferation of HUVEC cells and the death of certain TWEAK-sensitive tumor target cell lines. CD266 binds TWEAK (CD255) and can signal intracellularly by interactions with TRAF1, 2, and 3 cytoplasmic proteins leading to NF- κ B activation.
Specifications
| CD266 (TWEAK-R) | |
| Monoclonal | |
| 0.2mg/mL | |
| TWEAKR; TWEAK-R; TWEAK Receptor; TNFRSF12A; FGF-inducible 14; FN14 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| ITEM-1 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.