Learn More
CD25 Mouse anti-Rat, Brilliant Violet 421, Clone: OX-39, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565608
Description
The OX-39 monoclonal antibody specifically b inds to the α chain of the IL-2 R eceptor (IL-2R α , IL2RA) on activated T cells, T regulatory cells, and thymic and splenic dendritic cells. CD25 has also been detected on rat intestinal epithelial cells. It has been reported that OX-39 mAb weakly blocks binding of IL-2 to T-cell blasts and that it blocks IL-2 stimulated epithelial cell migration in an in vitro model of wound healing.
Specifications
| CD25 | |
| Monoclonal | |
| 0.2mg/mL | |
| Interleukin-2 receptor alpha chain; IL-2RA; IL-2Rα; Il2ra; IL-2R p55 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Rat | |
| IgG1 κ |
| Flow Cytometry | |
| OX-39 | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat T blasts from mixed-lymphocyte reactions | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.