missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22, Mouse, Clone: MEM-01, Abnova™
Mouse monoclonal antibody raised against native CD22.
Brand: Abnova MAB0908.100ug
Additional Details : Weight : 0.02000kg
Specifications
CD22 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
CD22 | |
CD22 | |
Native purified CD22 from Raji human Burkitt's lymphoma cell line. | |
RUO | |
933 | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
MEM-01 | |
Mouse monoclonal antibody raised against native CD22. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
FLJ22814/MGC130020/SIGLEC-2/SIGLEC2 | |
Mouse | |
100 μg | |
Primary | |
Human, Primate | |
Liquid |