missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2, Mouse, Clone: TP1/31, Abnova™
Mouse monoclonal antibody raised against native CD2.
Brand: Abnova MAB4958.100ug
This item is not returnable.
View return policy
Specifications
| CD2 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| CD2 | |
| CD2 | |
| Normal human peripheral blood lymphocytes. | |
| RUO | |
| 914 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence | |
| TP1/31 | |
| Mouse monoclonal antibody raised against native CD2. | |
| In buffer containing 1% BSA, pH 7.2 (0.09% sodium azide) | |
| SRBC/T11 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Sheep | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction