Learn More
CD14, Mouse, Clone: biG 14, Abnova™
Mouse monoclonal antibody raised against CD14.
Brand: Abnova MAB5655.100ug
Description
CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq
Specifications
| CD14 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (1:5000) The optimal working dilution should be determined by the end user. | |
| CD14 | |
| Mouse | |
| 100 μg | |
| Primary | |
| biG 14 represents an excellent marker for CD14. CD14 has been shown to be involved in the LPS-induced cell activation. This antigen is present as a membrane bound form and as soluble antigen, both involved in cell stimulation by LPS. The plasma protein LBP plays an important role in this LPS-CD14 mediated cell activation. Monocytes and PMN are activated by the LBP-LPS complex via membrane CD14. The CD14 glycoprotein, gp 55, is present on most monocytic and macrophages like cell types: monocytes, macrophages, Kupffer cells, pleural phagocytic cells and dendritic reticular cells. CD14 is also observed on granulocytes and activated or transformed B-cells. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids. The CD14 molecule has been reported to be a receptor for Endotoxin. | |
| Store at -20°C. For long term storage store at -80°C. After reconstitution with 100μL deionized water it is recommended to add 0.1% BSA. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a |
| ELISA, Flow Cytometry, Western Blot | |
| biG 14 | |
| Mouse monoclonal antibody raised against CD14. | |
| Lyophilized from PBS | |
| CD14 | |
| Monocytes of different species and immunoaffinity purified soluble human CD14. | |
| RUO | |
| 929 | |
| Bovine, Canine, Equine, Guinea Pig, Human, Pig | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.