Learn More
CD107b Mouse anti-Human, PE, Clone: H4B4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565306
Description
The H4B4 monoclonal antibody specifically binds to CD107b. CD107b is a heavily glycosylated, ∽120 kDa type I transmembrane protein that is also known as lysosomal-associated membrane protein 2 (LAMP-2/LAMP2), LAMPB, or LGP110. CD107b is expressed in the lysosomal and endosomal membranes of most cell types where it may play a role in maintaining membrane integrity. CD107b is also expressed on the surface of activated platelets and lymphocytes, including T lymphocytes, e.g., degranulating CD8+ cytotoxic T cells. CD107b serves as a ligand for selectins, including E-selectin, and mediates cellular adhesion. CD107a/ LAMP-1 and CD107b /LAMP-2 are carriers for poly-N-acetyllactosamines, and display the sialyl Le[x] antigen. CD107b is likewise expressed on some tumor cells and cell lines, including U937 and KG1a, and may facilitate tumor metastases.
Specifications
| CD107b | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Adult Adherent Peripheral Blood Cells | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry, Immunocytochemistry | |
| H4B4 | |
| LAMP-2; LAMP2; LAMPB; LGP110 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.