missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ccnb2, Mouse, Clone: X121.10, Abnova™
Mouse monoclonal antibody raised against full-length native ccnb2.
Brand: Abnova MAB6992.100ug
This item is not returnable.
View return policy
Specifications
| Ccnb2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| LOC397743 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Cyclin B2 (X121.10). WB predicted molecular weight is 48 KDa. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunoprecipitation, Western Blot | |
| X121.10 | |
| Mouse monoclonal antibody raised against full length native ccnb2. | |
| In PBS | |
| LOC397743 | |
| Native purified xenopus ccnb2. | |
| RUO | |
| 397743 | |
| Xenopus species | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction